Reagent for maintaining -SH groups in the reduced state, quantitatively reduces disulfides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for maintaining -SH groups in the reduced state, quantitatively reduces disulfides.
Solubility
Soluble in water (15.4 mg/ml), and methanol.
Notes
Air Sensitive, store away form air. Keep the container tightly sealed. Store away from reducing agent. Store under dry inert gas.
RUO – Research Use Only
Morgane Riou; David Stroebel; J Michael Edwardson; Pierre Paoletti. An alternating GluN1-2-1-2 subunit arrangement in mature NMDA receptors. Plosone. 2012, 7 (4), 35134 .
Yang-Rae Kim et. al. Highly sensitive gold nanoparticle-based colorimetric sensing of mercury(II) through simple ligand exchange reaction in aqueous media. ACS Applied Materials & Interfaces. 2010, 2 (1), 292-295.
Protective agent for preventing oxidation of thiol groups and reagent for reduction of disulfide groups in proteins: Biochemistry, 3, 480 (1964); 11, 2939 (1972); J. Biol. Chem., 243, 716 (1968); compare 1,4-Dithio-DL-threitol, A15797.