Thermo Scientific Chemicals

Cerium(IV) sulfate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A10142.0B
1000 g, Each
Thermo Scientific Chemicals

Cerium(IV) sulfate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A10142.0B
1000 g, Each
Quantity
Catalog number: A10142.0B
also known as A10142-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95838-16-7
IUPAC Name
λ⁴-cerium(4+) disulfate
Molecular Formula
CeO8S2
InChI Key
VZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Comment
Theoretical REO content (CeO2) dry wt. basis 51.80%
Elemental Analysis
Cerium 41.12 - 43.22% (Theory 42.17%), dry wt. basis
Appearance (Color)
Yellow
Form
crystalline powder
Assay (unspecified)
> 97.5% dry wt. basis

Description

Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text