Thermo Scientific Chemicals

2,3-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10144.18
50 g, Each
Thermo Scientific Chemicals

2,3-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10144.18
50 g, Each
Quantity
Catalog number: A10144.18
also known as A10144-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
303-38-8
IUPAC Name
2,3-dihydroxybenzoic acid
Molecular Formula
C7H6O4
InChI Key
GLDQAMYCGOIJDV-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC(O)=C1O
Appearance (Color)
Pale cream to pale brown or pale pink
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%

Description

Dihydroxybenzoic acids are used as intermediates for pharmaceuticals and other organic synthesis. Used as matrix for ionization of peptides, proteins and carbohydrates. In industrial field used for resins, polyesters, plasticizers, dyestuffs, preservatives, and rubber chemicals. It is also used as is a selected phenol as MMP (Matrix Metalloproteinase) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dihydroxybenzoic acids are used as intermediates for pharmaceuticals and other organic synthesis. Used as matrix for ionization of peptides, proteins and carbohydrates. In industrial field used for resins, polyesters, plasticizers, dyestuffs, preservatives, and rubber chemicals. It is also used as is a selected phenol as MMP (Matrix Metalloproteinase) inhibitor.

Solubility
Soluble in water, methanol and dimethyl sulfoxide.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text