Thermo Scientific Chemicals

2-Iodoanisole, 99%, Thermo Scientific Chemicals

Catalog number: A10150.36
500 g, Each
Thermo Scientific Chemicals

2-Iodoanisole, 99%, Thermo Scientific Chemicals

Catalog number: A10150.36
500 g, Each
Quantity
Catalog number: A10150.36
also known as A10150-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
529-28-2
IUPAC Name
1-iodo-2-methoxybenzene
Molecular Formula
C7H7IO
InChI Key
DVQWNQBEUKXONL-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1I
Appearance (Color)
Clear pale yellow to yellow
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.6210-1.6240 @ 20?C

Description

2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.

Solubility
Miscible with alcohol, chloroform and diethyl ether. Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text