Thermo Scientific Chemicals

Diethoxymethylsilane, 96%, Thermo Scientific Chemicals

Catalog number: A10153.09
10 g, Each
Thermo Scientific Chemicals

Diethoxymethylsilane, 96%, Thermo Scientific Chemicals

Catalog number: A10153.09
10 g, Each
Quantity
Catalog number: A10153.09
also known as A10153-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2031-62-1
IUPAC Name
diethoxy(methyl)silyl
Molecular Formula
C5H13O2Si
InChI Key
GAURFLBIDLSLQU-UHFFFAOYSA-N
SMILES
CCO[Si](C)OCC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Refractive Index
1.3730-1.3790 @ 20?C

Description

Diethoxymethylsilane is used in the preparation of N,N'-methylsilanediyl-bis-phthalimide by reacting with phthalimide. It is used as a reagent in the selective reduction of carbonyl compounds, hydorosilylation of alkenes, tandem reductive aldol reaction and rhodium-catalyszed silylcarbocyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethoxymethylsilane is used in the preparation of N,N′-methylsilanediyl-bis-phthalimide by reacting with phthalimide. It is used as a reagent in the selective reduction of carbonyl compounds, hydorosilylation of alkenes, tandem reductive aldol reaction and rhodium-catalyszed silylcarbocyclization.

Solubility
Miscible with water, 1,4-dioxane, tetrahydrofuran, acetonitrile, dichloromethane, chloroform, dimethyl sulfoxide, hexamethylphosphoric triamide and toluene. Insoluble in diethyl ether and hexane.

Notes
Incompatible with oxidizing agents, acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text