Thermo Scientific Chemicals

tert-Butylacetyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A10162.22
100 g, Each
Thermo Scientific Chemicals

tert-Butylacetyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A10162.22
100 g, Each
Quantity
Catalog number: A10162.22
also known as A10162-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7065-46-5
IUPAC Name
3,3-dimethylbutanoyl chloride
Molecular Formula
C6H11ClO
InChI Key
BUTKIHRNYUEGKB-UHFFFAOYSA-N
SMILES
CC(C)(C)CC(Cl)=O
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.4205-1.4255 @ 20?C
Assay (GC)
≥98.0%

Description

tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

Solubility
Reacts with water.

Notes
Moisture Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text