Thermo Scientific Chemicals

3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: A10167.0B
1000 g, Each
Thermo Scientific Chemicals

3,4-Dimethoxybenzoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: A10167.0B
1000 g, Each
Quantity
Catalog number: A10167.0B
also known as A10167-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-07-2
IUPAC Name
3,4-dimethoxybenzoic acid
Molecular Formula
C9H10O4
InChI Key
DAUAQNGYDSHRET-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1OC)C(O)=O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥99.0 to ≤101.0%
Melting Point (clear melt)
179.0-186.0?C
Form
Powder
Assay (Silylated GC)
≥99.0%

Description

3,4-Dimethoxybenzoic acid plays an important role in producing antibiotics and various dyes. They are used as intermediates for pharmaceuticals (especially for antipyretic analgesic, antirheumatism) and other organic synthesis. It is used as matrix for ionization of peptides, proteins and carbohydrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxybenzoic acid plays an important role in producing antibiotics and various dyes. They are used as intermediates for pharmaceuticals (especially for antipyretic analgesic, antirheumatism) and other organic synthesis. It is used as matrix for ionization of peptides, proteins and carbohydrates.

Solubility
Soluble in water (0.5 mg/ml at 15°C), chloroform, methanol, and 95% ethanol (50 mg/ml).

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text