Thermo Scientific Chemicals

Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A10168.22
100 g, Each
Thermo Scientific Chemicals

Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A10168.22
100 g, Each
Quantity
Catalog number: A10168.22
also known as A10168-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15244-35-6
IUPAC Name
cadmium(2+) sulfate
Molecular Formula
CdO4S
InChI Key
QCUOBSQYDGUHHT-UHFFFAOYSA-L
SMILES
[Cd++].[O-]S([O-])(=O)=O
Appearance (Color)
White
Complexometric Titration
>97.5 to <102.5% (dry wt. basis)
Comment
Waters of hydration ca 3/8
Form
Crystalline powder

Description

Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

Solubility
Soluble in water (1130g/L ).

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text