Thermo Scientific Chemicals

3-Chloro-4-fluorophenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A10175.14
25 g, Each
Thermo Scientific Chemicals

3-Chloro-4-fluorophenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A10175.14
25 g, Each
Quantity
Catalog number: A10175.14
also known as A10175-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
175135-74-7
IUPAC Name
(3-chloro-4-fluorophenyl)hydrazine
Molecular Formula
C6H6ClFN2
InChI Key
DUORBNTWCCJANU-UHFFFAOYSA-N
SMILES
NNC1=CC=C(F)C(Cl)=C1
Appearance (Color)
Pale cream to pale brown or pale pink
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text