Calcium fluoride, 97%, Thermo Scientific Chemicals
Calcium fluoride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium fluoride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A10176.30
also known as A10176-30
Price (USD)/ Each
26.65
Online exclusive
29.00 
Save 2.35 (8%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
26.65
Online exclusive
29.00 
Save 2.35 (8%)
Add to cart
Calcium fluoride, 97%, Thermo Scientific Chemicals
Catalog numberA10176.30
Price (USD)/ Each
26.65
Online exclusive
29.00 
Save 2.35 (8%)
-
Add to cart
Chemical Identifiers
CAS7789-75-5
IUPAC Namecalcium difluoride
Molecular FormulaCaF2
InChI KeyWUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES[F-].[F-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
FormPowder
Assay from Supplier's CofA≥96.0%
Loss on Drying<2.5%
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.

Solubility
Soluble in ammonium salts. Soluble in water (slightly).

Notes
Store at room temperature. Separated from mineral acids. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
RUO – Research Use Only

General References:

  1. Tao Pan; Ryan T. Kelly; Matthew C. Asplund; Adam T. Woolley. Fabrication of calcium fluoride capillary electrophoresis microdevices for on-chip infrared detection. Journal of Chromatography A. 2004, 1027 231-235.
  2. H G McCann. The solubility of fluorapatite and its relationship to that of calcium fluoride. Archives of Oral Biology. 1968, 13 (8), 987-1001.
  3. In combination with KF, is a useful fluorinating agent for conversion of other alkyl halides to fluorides: J. Chem. Soc., Chem. Commun., 793 (1986).