Thermo Scientific Chemicals

L-Alanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A10178.06
5 g, Each
Thermo Scientific Chemicals

L-Alanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A10178.06
5 g, Each
Quantity
Catalog number: A10178.06
also known as A10178-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2491-20-5
IUPAC Name
methyl (2S)-2-aminopropanoate hydrochloride
Molecular Formula
C4H10ClNO2
InChI Key
IYUKFAFDFHZKPI-DOIAFFBONA-N
SMILES
Cl.COC(=O)[C@H](C)N
Assay (Titration ex Chloride)
≥98.5%
Appearance (Color)
White to pale cream
Identification (FTIR)
Conforms
Melting Point (clear melt)
104-110?C
Optical Rotation
+7.0? ? 1.0? (c=1.6 in methanol)

Description

L-Alanine methyl ester hydrochloride is used to make in-vivo measurement of glucose and alanine metabolism in studies of patients with diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solubility
Soluble in Water (100 mg/ml).

Notes
Hygroscopic store under inert gas Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text