Thermo Scientific Chemicals

n-Propyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals

Catalog number: A10180.30
250 g, Each
Thermo Scientific Chemicals

n-Propyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals

Catalog number: A10180.30
250 g, Each
Quantity
Catalog number: A10180.30
also known as A10180-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-13-3
IUPAC Name
propyl 4-hydroxybenzoate
Molecular Formula
C10H12O3
InChI Key
QELSKZZBTMNZEB-UHFFFAOYSA-N
SMILES
CCCOC(=O)C1=CC=C(O)C=C1
Appearance (Color)
White to pale cream
Melting Point (clear melt)
95-100?C
Form
Powder or crystalline powder
Identification (FTIR)
Conforms
Assay (GC)
≥99.0%

Description

n-Propyl 4-hydroxybenzoate is used as antimicrobial preservatives in pharmaceuticals, foods, cosmetics and shampoos. It is widely used as preservatives in personal care products like creams, lotions and bath products. Further, it is used as a food additive. Its derivative, sodium propyl p-hydroxybenzoate is used as an anti-fungal preservation agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Propyl 4-hydroxybenzoate is used as antimicrobial preservatives in pharmaceuticals, foods, cosmetics and shampoos. It is widely used as preservatives in personal care products like creams, lotions and bath products. Further, it is used as a food additive. Its derivative, sodium propyl p-hydroxybenzoate is used as an anti-fungal preservation agent.

Solubility
Soluble in propylene glycol, ethanol, ether, acetone, methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and alkali metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text