Thermo Scientific Chemicals

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%, Thermo Scientific Chemicals

Catalog number: A10181.22
100 g, Each
Thermo Scientific Chemicals

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%, Thermo Scientific Chemicals

Catalog number: A10181.22
100 g, Each
Quantity
Catalog number: A10181.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6001-64-5
IUPAC Name
bis(1,1,1-trichloro-2-methylpropan-2-ol) hydrate
Molecular Formula
C8H16Cl6O3
InChI Key
WRWLCXJYIMRJIN-UHFFFAOYSA-N
SMILES
O.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl
Water Content (Karl Fischer Titration)
3.90-6.63% (0.5 waters, 4.83%)
Form
Crystals or Crystalline powder
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Appearance (Color)
White

Description

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate converts benzisoxazole to α-aryloxyisobutyric acid. It forms eutectic with dimethyl sulfone, which is the most suitable media for freeze-drying due to its high solubilizing ability and a good rate of solvent removal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate converts benzisoxazole to α-aryloxyisobutyric acid. It forms eutectic with dimethyl sulfone, which is the most suitable media for freeze-drying due to its high solubilizing ability and a good rate of solvent removal.

Solubility
Soluble in ethanol, ether, chloroform, and glycerol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text