4,4'-Bipyridine, 98%, Thermo Scientific Chemicals
4,4'-Bipyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Bipyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A10187.14
also known as A10187-14
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
Add to cart
4,4'-Bipyridine, 98%, Thermo Scientific Chemicals
Catalog numberA10187.14
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS553-26-4
IUPAC Name4,4'-bipyridine
Molecular FormulaC10H8N2
InChI KeyMWVTWFVJZLCBMC-UHFFFAOYSA-N
SMILESC1=CC(=CC=N1)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder
Water Content (Karl Fischer Titration)≤2.0%
Assay (GC)≥97.5%
Appearance (Color)White to cream or pale yellow or pale brown
Identification (FTIR)Conforms
View more
4,4'-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N'-dimethyl-4,4'-bipyridinium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N′-dimethyl-4,4′-bipyridinium.

Solubility
Soluble in ethanol, ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Ohmura, T.; Morimasa, Y.; Suginome, M. Organocatalytic Diboration Involving Reductive Addition of a Boron-Boron σ-Bond to 4,4'-Bipyridine. J. Am. Chem. Soc. 2015, 137 (8), 2852-2855.
  2. Aniola, M.; Katrusiak, A. Conformational Conversion of 4,4'-Bipyridinium in a Hidden High-Pressure Phase. Cryst. Growth Des. 2015, 15 (2), 764-770.