Thermo Scientific Chemicals

Benzoin, 99%, Thermo Scientific Chemicals

Catalog number: A10188.30
250 g, Each
Thermo Scientific Chemicals

Benzoin, 99%, Thermo Scientific Chemicals

Catalog number: A10188.30
250 g, Each
Quantity
Catalog number: A10188.30
also known as A10188-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-53-9
Melting Point (clear melt)
129-137?C
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Appearance (Color)
White to cream or pale yellow

Description

Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.

Solubility
Soluble in chlorine. Slightly soluble in water, ethanol and ether.

Notes
Moisture sensitive. Incompatible with acids, acid anhydrides, acid chlorides and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text