Thermo Scientific Chemicals

Ammonium thiosulfate, 96%, Thermo Scientific Chemicals

Catalog number: A10190.22
100 g, Each
Thermo Scientific Chemicals

Ammonium thiosulfate, 96%, Thermo Scientific Chemicals

Catalog number: A10190.22
100 g, Each
Quantity
Catalog number: A10190.22
also known as A10190-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-18-8
IUPAC Name
dihydroxy-1λ⁶-disulfen-1-one diamine
Molecular Formula
H8N2O3S2
InChI Key
XYXNTHIYBIDHGM-UHFFFAOYSA-N
SMILES
N.N.OS(O)(=O)=S
Assay from Supplier's CofA
≥95.0%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused/lumpy solid or chunks or flakes

Description

Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.

Solubility
Soluble in water, slightly soluble in acetone. Insoluble in ethanol and diethyl ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text