The ATTO-TAG FQ reagent allows the ultrasensitive detection of primary amines. This reagent is available as a standalone product or as a component of an amine-derivatization kit (A-2334). Because ATTO-TAG FQ is nonfluorescent in the unreacted state and reacts specifically with amines to form highly fluorescent conjugates, it is particularly well-suited to the derivatization of peptides, amino acids, aminated sugars and other amine-containing molecules for detection by capillary electrophoresis with laser-induced fluorescence (LIF). Recently, researchers described a simple method in which the ATTO-TAG FQ reagent was used to derivatize very dilute solutions (10-8 M) of peptides.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Reactivity
Amine
Reactive Moiety
Aldehyde
Shipping Condition
Room Temperature
Form
Solid
Product Line
ATTO-TAG™
Quantity
10 mg
For Use With (Application)
Detection of primary amines
Target
Primary Amine
Type
Derivatization Kits and Reagents
Label or Dye
ATTO-TAG FQ
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.