Thermo Scientific Chemicals

L-Proline, 99%, Thermo Scientific Chemicals

Catalog number: A10199.14
25 g, Each
Thermo Scientific Chemicals

L-Proline, 99%, Thermo Scientific Chemicals

Catalog number: A10199.14
25 g, Each
Quantity
Catalog number: A10199.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
147-85-3
IUPAC Name
pyrrolidine-2-carboxylic acid
Molecular Formula
C5H9NO2
InChI Key
ONIBWKKTOPOVIA-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCN1
Form
Crystals or powder or crystalline powder
Optical Rotation
-84 ? 3? (c= 1 in water)
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in collagen. It is an active component of collagen and involved in the proper functioning of joints and tendons. It finds uses in pharmaceutical, biotechnological applications due to its osmoprotectant property. Further, it is used with ninhydrin in the chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in collagen. It is an active component of collagen and involved in the proper functioning of joints and tendons. It finds uses in pharmaceutical, biotechnological applications due to its osmoprotectant property. Further, it is used with ninhydrin in the chromatography.

Solubility
Soluble in water and ethanol. Insoluble in ether, butanol and isopropanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text