4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A10208.18
also known as A10208-18
Price (USD)/ Each
208.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
208.00
Add to cart
4-Fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
Catalog numberA10208.18
Price (USD)/ Each
208.00
-
Add to cart
Chemical Identifiers
CAS459-46-1
IUPAC Name1-(bromomethyl)-4-fluorobenzene
Molecular FormulaC7H6BrF
InChI KeyNVNPLEPBDPJYRZ-UHFFFAOYSA-N
SMILESFC1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.5455-1.5505 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only