Thermo Scientific Chemicals

1-Adamantanol, 99%, Thermo Scientific Chemicals

Catalog number: A10209.09
10 g, Each
Thermo Scientific Chemicals

1-Adamantanol, 99%, Thermo Scientific Chemicals

Catalog number: A10209.09
10 g, Each
Quantity
Catalog number: A10209.09
also known as A10209-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
768-95-6
IUPAC Name
adamantan-1-ol
Molecular Formula
C10H16O
InChI Key
VLLNJDMHDJRNFK-UHFFFAOYSA-N
SMILES
OC12CC3CC(CC(C3)C1)C2
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%

Description

1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.

Solubility
Soluble in ethanol , methanol, and diethyl ether. Partly miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text