Thermo Scientific Chemicals

Tetraethylammonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: A10211.30
250 g, Each
Thermo Scientific Chemicals

Tetraethylammonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: A10211.30
250 g, Each
Quantity
Catalog number: A10211.30
also known as A10211-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
429-06-1
IUPAC Name
tetraethylazanium; tetrafluoroboranuide
Molecular Formula
C8H20BF4N
InChI Key
XJRAKUDXACGCHA-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CC[N+](CC)(CC)CC
Assay from Suppliers CofA
Boron: ≥ 4.9% (U.S. specification)
Water Content (Karl Fischer Titration)
≤0.8% (non-U.S. specification)
Assay (unspecified)
≥98.5% by C & N EA (non-U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Appearance (Color)
White

Description

Tetraethylammonium tetrafluoroborate is used as a phase transfer catalyst in organic synthesis. It finds application as an electrolyte additive in the capillary electrophoretic separation of polyphenols found in grape seed extracts. It is also used to prepare other tetrabutylammonium salts in aqueous solutions. It is involved in the synthesis of conducting poly(thiophenes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium tetrafluoroborate is used as a phase transfer catalyst in organic synthesis. It finds application as an electrolyte additive in the capillary electrophoretic separation of polyphenols found in grape seed extracts. It is also used to prepare other tetrabutylammonium salts in aqueous solutions. It is involved in the synthesis of conducting poly(thiophenes).

Solubility
Soluble in water, alcohol and acetonitrile.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text