Thermo Scientific Chemicals

2-Acetylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10216.30
250 g, Each
Thermo Scientific Chemicals

2-Acetylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10216.30
250 g, Each
Quantity
Catalog number: A10216.30
also known as A10216-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
577-56-0
IUPAC Name
2-acetylbenzoic acid
Molecular Formula
C9H8O3
InChI Key
QDAWXRKTSATEOP-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC=C1C(O)=O
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream or pale yellow to pale orange
Melting Point (clear melt)
114.0-120.0?C

Description

2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.

Solubility
Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.

Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text