Thermo Scientific Chemicals

Phthalan, 98%, Thermo Scientific Chemicals

Catalog number: A10217.06
5 g, Each
Thermo Scientific Chemicals

Phthalan, 98%, Thermo Scientific Chemicals

Catalog number: A10217.06
5 g, Each
Quantity
Catalog number: A10217.06
also known as A10217-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
496-14-0
IUPAC Name
1,3-dihydro-2-benzofuran
Molecular Formula
C8H8O
InChI Key
SFLGSKRGOWRGBR-UHFFFAOYSA-N
SMILES
C1OCC2=CC=CC=C12
Refractive Index
1.5440-1.5480 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4'-Di-tert-butyl­biphenyl­, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4′-Di-tert-butyl­biphenyl­, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.

Solubility
Low aqueous solubility.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text