Thermo Scientific Chemicals

4-Bromo-2-fluoroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A10228.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-2-fluoroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A10228.14
25 g, Each
Quantity
Catalog number: A10228.14
also known as A10228-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
367-24-8
IUPAC Name
4-bromo-2-fluoroaniline
Molecular Formula
C6H5BrFN
InChI Key
GZRMNMGWNKSANY-UHFFFAOYSA-N
SMILES
NC1=CC=C(Br)C=C1F
Identification (FTIR)
Conforms
Melting Point (clear melt)
36.0-43.0?C
Appearance (Color)
White to pale cream to cream to brown to pale gray
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (GC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text