Thermo Scientific Chemicals

L-Tryptophan, 99%

Catalog number: A10230.36
500 g, Each
Thermo Scientific Chemicals

L-Tryptophan, 99%

Catalog number: A10230.36
500 g, Each
Quantity
Catalog number: A10230.36
also known as A10230-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73-22-3
IUPAC Name
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
Molecular Formula
C11H12N2O2
InChI Key
QIVBCDIJIAJPQS-VIFPVBQESA-N
SMILES
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
Optical Rotation
-31.5? ? 2? (c= 1 in Water)
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.5% to ≤101.5% (UK Sourced Material)
Assay from Supplier's CofA
≥98.5% (US Sourced Material)
Comment
Material Sourced in UK and US

Description

L-tryptophan is an essential amino acid which is necessary for normal growth in infants and for nitrogen balance in adults. It acts as a natural dietary supplement and used as an antidepressant, anxiolytic and sleep aid. It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-tryptophan is an essential amino acid which is necessary for normal growth in infants and for nitrogen balance in adults. It acts as a natural dietary supplement and used as an antidepressant, anxiolytic and sleep aid. It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.

Solubility
Soluble in water, ammonia, hot alcohol, alkali hydroxides and dilute acids. Slightly soluble in acetic acid and ethanol. Insoluble in chloroform and ethyl ether.

Notes
Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text