Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Ready to use in just 90 minutes, our protein labeling kits include an easy-to-use, pre-packed spin column for rapid dye removal and typical recovery greater than 85%. Each kit contains enough reagent for 3–5 protein conjugation reactions. Our kits provide better results due to lower background fluorescence, less nonspecific binding, and easier workflows for protein conjugation with 16 different fluorophores.
Description | Alexa Fluor™ 488 Protein Labeling Kit |
Product Type | Labeling Kit |
Detection Method | Fluorescence |
Excitation/Emission | 494/519 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 488 |
Color | Green |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
Description | Alexa Fluor™ 350 Protein Labeling Kit |
Product Type | Labeling Kit |
Detection Method | Fluorescence |
Excitation/Emission | 346/442 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 350 |
Color | Blue |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
Description | Alexa Fluor™ 532 Protein Labeling Kit |
Product Type | Labeling Kit |
Quantity | 3 Reactions |
Detection Method | Fluorescence |
Excitation/Emission | 530/554 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 532 |
Color | Yellow |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
Description | Alexa Fluor™ 546 Protein Labeling Kit |
Product Type | Labeling Kit |
Detection Method | Fluorescence |
Excitation/Emission | 554/570 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 546 |
Color | Orange |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
Description | Alexa Fluor™ 555 Protein Labeling Kit |
Product Type | Labeling Kit |
Detection Method | Fluorescence |
Excitation/Emission | 553/565 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 555 |
Color | Orange |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
Description | Alexa Fluor™ 568 Protein Labeling Kit |
Product Type | Labeling Kit |
Detection Method | Fluorescence |
Excitation/Emission | 577/603 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 568 |
Color | Orange-Red |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
Description | Alexa Fluor™ 594 Protein Labeling Kit |
Product Type | Labeling Kit |
Detection Method | Fluorescence |
Excitation/Emission | 590/617 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 594 |
Color | Red |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
Description | Alexa Fluor™ 633 Protein Labeling Kit |
Product Type | Labeling Kit |
Detection Method | Fluorescence |
Excitation/Emission | 633/647 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 633 |
Color | Far-Red |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
Description | Alexa Fluor™ 647 Protein Labeling Kit |
Product Type | Labeling Kit |
Detection Method | Fluorescence |
Excitation/Emission | 650/668 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 647 |
Color | Far-Red |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
Description | Alexa Fluor™ 660 Protein Labeling Kit |
Product Type | Labeling Kit |
Detection Method | Fluorescence |
Excitation/Emission | 663/690 |
Labeling Method | Conjugation-Based |
Labeling Scale | 1 mg |
Shipping Condition | Room Temperature |
Product Line | Alexa Fluor™ |
Chemical Reactivity | Amine |
Solubility | DMSO (Dimethylsulfoxide) |
Label or Dye | Alexa Fluor 660 |
Color | Far-Red |
Labeling Target | Antibodies, Proteins |
Label Type | Alexa Fluor Dyes |
Unit Size | 1 kit |
產品號碼 | 規格 | Unit Size | Description | 價格 (HKD) | 供應狀況 | 數量 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A10235 | 1 kit | Alexa Fluor™ 488 Protein Labeling Kit | 價格 5,833.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A10170 | 1 kit | Alexa Fluor™ 350 Protein Labeling Kit | 價格 5,846.00 線上優惠 4,678.00 (31-Aug-2024 結束) 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A10236 | 1 kit | Alexa Fluor™ 532 Protein Labeling Kit | 價格 5,533.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A10237 | 1 kit | Alexa Fluor™ 546 Protein Labeling Kit | 價格 5,844.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A20174 | 1 kit | Alexa Fluor™ 555 Protein Labeling Kit | 價格 5,848.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A10238 | 1 kit | Alexa Fluor™ 568 Protein Labeling Kit | 價格 5,846.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A10239 | 1 kit | Alexa Fluor™ 594 Protein Labeling Kit | 價格 5,847.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A20170 | 1 kit | Alexa Fluor™ 633 Protein Labeling Kit | 價格 5,846.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A20173 | 1 kit | Alexa Fluor™ 647 Protein Labeling Kit | 價格 5,833.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A20171 | 1 kit | Alexa Fluor™ 660 Protein Labeling Kit | 價格 5,825.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Each protein labeling kit contains everything you need to perform 3-5 separate labeling reactions and purify the resulting conjugates. The reactive dye has either a succinimidyl ester (SE) or a tetrafluorophenyl (TFP) ester moiety that reacts efficiently with primary amines of proteins to form stable dye–protein conjugates. Each of the vials of reactive dye provided in the kit is sufficient for labeling 1 mg of a variety of purified proteins, including growth factors, cytokines, nanobodies, enzymes, cell-adhesion molecules, and antibodies.
Direct labeling with fluorophores allows multiple primary antibodies of the same isotype (derived from the same species) to be used in the same experiment. Stabilizing proteins such as BSA should be removed from the sample before labeling.
The different protein labeling kits
For labeling smaller amounts of antibodies (∼100 μg), we recommend our antibody labeling kits. Please review the protein labeling kit user manual for more in depth information, protocols, molecular weights, and degree of labeling for each dye.