Polyethylene powder, low density, 500 micron, Thermo Scientific Chemicals
Polyethylene powder, low density, 500 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyethylene powder, low density, 500 micron, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number A10239.36
also known as A10239-36
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
Add to cart
Polyethylene powder, low density, 500 micron, Thermo Scientific Chemicals
Catalog numberA10239.36
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9002-88-4
Molecular Formula(C2H4)n
SMILES*-CC-*
MDL NumberMFCD00084423
SpecificationsSpecification SheetSpecification Sheet
Density @20°C(Resin): 0.9100-0.9600 g/ml
FormPowder
Particle Sizeapprox. 500 micron
Appearance (Color)White
CommentMelt flow index: 17.5-23.5 g/10min (190?C/2.16kg)
Polyethylene powder is used as an additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of ultra-high-molecular-weight polyethylene (UHMWPE). It is widely used for manufacturing various containers, trays, dispensing bottles, plastic wraps, plastic bags for computer components and various molded laboratory equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only