Thermo Scientific Chemicals

Sodium 3-nitrobenzenesulfonate, 98%, water <2%, Thermo Scientific Chemicals

Catalog number: A10244.22
100 g, Each
Thermo Scientific Chemicals

Sodium 3-nitrobenzenesulfonate, 98%, water <2%, Thermo Scientific Chemicals

Catalog number: A10244.22
100 g, Each
Quantity
Catalog number: A10244.22
also known as A10244-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-68-4
IUPAC Name
sodium 3-nitrobenzene-1-sulfonate
Molecular Formula
C6H4NNaO5S
InChI Key
LJRGBERXYNQPJI-UHFFFAOYSA-M
SMILES
[Na+].[O-][N+](=O)C1=CC=CC(=C1)S([O-])(=O)=O
Appearance (Color)
White to cream or pale yellow to brown
Form
Powder
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤2.0%

Description

Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.

Solubility
Soluble in water 200 g/L (20C).

Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text