Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.
Solubility
Soluble in water 200 g/L (20C).
Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Rong-Rong Li; Hua-Ding Liang; Chang-Hua Ge; Cai-Ping Lin; Yu-Jing You; and Bo Zhao. Equilibrium Solubility of Sodium 3-Nitrobenzenesulfonate in Binary NaCl + H2O, Na2SO4 + H2O, and C2H5OH + H2O Solvent Mixtures at Elevated Temperatures from (273.15 to 323.15) K. J. Chem. Eng. Data. 2012, 57,(12), 3434-3437.
Dwight Macdonald et. al. Discovery of a substituted 8-arylquinoline series of PDE4 inhibitors: structure-activity relationship, optimization, and identification of a highly potent, well tolerated, PDE4 inhibitor. Bioorganic & Medicinal Chemistry Letters. 2005, 15,(23), 5241-5246.