Thermo Scientific Chemicals

4-n-Pentylbenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10248.06
5 g, Each
Thermo Scientific Chemicals

4-n-Pentylbenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10248.06
5 g, Each
Quantity
Catalog number: A10248.06
also known as A10248-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
49763-65-7
IUPAC Name
4-pentylbenzoyl chloride
Molecular Formula
C12H15ClO
InChI Key
FBBRKYLXMNQFQU-UHFFFAOYSA-N
SMILES
CCCCCC1=CC=C(C=C1)C(Cl)=O
Assay (GC)
>97.5%
Refractive Index
1.5280-1.5320 @ 20?C
Comment
Clear colorless to pale yellow
Form
Liquid

Description

4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text