4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.
RUO – Research Use Only
Rama Mukherjee; Manu Jaggi; Praveen Rajendran; Sanjay K Srivastava; Mohammad J A Siddiqui; Anand Vardhan; Anand C Burman. Synthesis of 3-O-acyl/3-benzylidene/3-hydrazone/3-hydrazine/17-carboxyacryloyl ester derivatives of betulinic acid as anti-angiogenic agents. Bioorganic & Medicinal Chemistry Letters. 2004, 14 (12), 3169-3172.