Thermo Scientific Chemicals

Tetra-n-butylammonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A10249.36
500 g, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A10249.36
500 g, Each
Quantity
Catalog number: A10249.36
also known as A10249-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1643-19-2
IUPAC Name
tetrabutylazanium bromide
Molecular Formula
C16H36BrN
InChI Key
JRMUNVKIHCOMHV-UHFFFAOYSA-M
SMILES
[Br-].CCCC[N+](CCCC)(CCCC)CCCC
Form
Crystals or powder or crystalline powder or fused solid
Water Content (Karl Fischer Titration)
≤1%
Appearance (Color)
White to cream
Assay (Titration ex Bromide)
≥98.0 to ≤102.0% (UK Sourced Material)
Comment
Material Sourced in UK and US

Description

Phase transfer catalyst Tetra-n-butylammonium bromide is used as a phase transfer catalyst for the preparation of lactones by cyclization of potassium salt of μ-bromocarboxylic acids and beta-lactones from beta-bromoacid by cyclization. It is also involved in the Curtius rearrangement, N-alkylation of aromatic carboxamides. It plays an important role as an ionic liquid in the regioselective O-alkylation reactions. It is also used in the preparation of polymer solar cells and single component green-light emitting electrochemical cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phase transfer catalyst Tetra-n-butylammonium bromide is used as a phase transfer catalyst for the preparation of lactones by cyclization of potassium salt of µ-bromocarboxylic acids and beta-lactones from beta-bromoacid by cyclization. It is also involved in the Curtius rearrangement, N-alkylation of aromatic carboxamides. It plays an important role as an ionic liquid in the regioselective O-alkylation reactions. It is also used in the preparation of polymer solar cells and single component green-light emitting electrochemical cells.

Solubility
Soluble in water and ethanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text