3-Methylcyclopentane-1,2-dione, 98+%, Thermo Scientific Chemicals
3-Methylcyclopentane-1,2-dione, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methylcyclopentane-1,2-dione, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
100 g
Catalog number A10255.18
also known as A10255-18
Price (USD)/ Each
91.60
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
91.60
Add to cart
3-Methylcyclopentane-1,2-dione, 98+%, Thermo Scientific Chemicals
Catalog numberA10255.18
Price (USD)/ Each
91.60
-
Add to cart
Chemical Identifiers
CAS765-70-8
IUPAC Name3-methylcyclopentane-1,2-dione
Molecular FormulaC6H8O2
InChI KeyOACYKCIZDVVNJL-UHFFFAOYNA-N
SMILESCC1CCC(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)White to cream
Melting Point (clear melt)102.0-108.0?C
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only