2-Ketoglutaric acid is used as an intermediate in the kreb’s cycle catalyzed by glutamate dehydrogenase enzyme on glutamate. It is also used in dietary supplements to improve protein synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ketoglutaric acid is used as an intermediate in the kreb’s cycle catalyzed by glutamate dehydrogenase enzyme on glutamate. It is also used in dietary supplements to improve protein synthesis.
Solubility
Soluble in water, methanol and dimethyl sulfoxide.
Notes
Store away from heat store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
T. Garcia-Rodriguez; A.M. GuTierrez-Navarro; R. Garcia; J.Perez Silva. Indole acetic acid production by Rhizobium: Effect of 2-ketoglutaric acid. Soil Biol. Biochem. 1982, 14,(2), 153-155.
F. Rocchiccioli; J. P. Leroux; and P. H. Cartier. Microdetermination of 2-ketoglutaric acid in plasma and cerebrospinal fluid by capillary gas chromatography mass spectrometry; application to pediatrics. Biological Mass Spectrometry. 1984, 11,(1), 24-28.