Thermo Scientific Chemicals

Methyl tetradecanoate, 99%, Thermo Scientific Chemicals

Catalog number: A10257.18
50 g, Each
Thermo Scientific Chemicals

Methyl tetradecanoate, 99%, Thermo Scientific Chemicals

Catalog number: A10257.18
50 g, Each
Quantity
Catalog number: A10257.18
also known as A10257-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-10-7
IUPAC Name
methyl tetradecanoate
Molecular Formula
C15H30O2
InChI Key
ZAZKJZBWRNNLDS-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCC(=O)OC
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Refractive Index
1.4345-1.4375 @ 20?C
Assay (GC)
≥98.5%

Description

Methyl tetradecanoate is used as a component in the preparation of a high-density biodiesel. Also used as a niosome medium in which an anti-cancer drug could be transported within.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl tetradecanoate is used as a component in the preparation of a high-density biodiesel. Also used as a niosome medium in which an anti-cancer drug could be transported within.

Solubility
Soluble in chloroform and methanol. React with water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text