Methyl tetradecanoate is used as a component in the preparation of a high-density biodiesel. Also used as a niosome medium in which an anti-cancer drug could be transported within.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl tetradecanoate is used as a component in the preparation of a high-density biodiesel. Also used as a niosome medium in which an anti-cancer drug could be transported within.
Solubility
Soluble in chloroform and methanol. React with water.
Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Balasubramanian Narasimhan et. al. Design, synthesis, antibacterial, and QSAR studies of myristic acid derivatives. Bioorg. Med. Chem. Lett. 2006, 16,(11), 3434-3437.