Thermo Scientific Chemicals

2-Hydroxy-5-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A10264.22
100 g, Each
Thermo Scientific Chemicals

2-Hydroxy-5-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A10264.22
100 g, Each
Quantity
Catalog number: A10264.22
also known as A10264-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-51-8
IUPAC Name
2-hydroxy-5-nitrobenzaldehyde
Molecular Formula
C7H5NO4
InChI Key
IHFRMUGEILMHNU-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1C=O)[N+]([O-])=O
Appearance (Color)
Pale yellow to yellow to orange
Assay (GC)
≥97.5%
Form
Powder
Melting Point (clear melt)
122.0-129.0?C

Description

2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

Solubility
Soluble in water (partly).

Notes
Air & Light Sensitive,. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text