Thermo Scientific Chemicals

Lead(II) sulfide, Pb 82% min., Thermo Scientific Chemicals

Catalog number: A10268.0E
2500 g, Each
Thermo Scientific Chemicals

Lead(II) sulfide, Pb 82% min., Thermo Scientific Chemicals

Catalog number: A10268.0E
2500 g, Each
Quantity
Catalog number: A10268.0E
also known as A10268-0E
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1314-87-0
IUPAC Name
λ²-lead(2+) sulfanediide
Molecular Formula
PbS
InChI Key
FILXVLQXZYXBSJ-UHFFFAOYSA-N
SMILES
[S--].[Pb++]
Appearance (Color)
Grey to purple
Form
Powder
Elemental analysis by ICP
Pb content ≥78%

Description

Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.

Solubility
Soluble in water (0.00086g/L) and acid. Insoluble in alcohol, and potassium hydroxide.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text