Thermo Scientific Chemicals

5-Chloro-2-pentanone, 94%, Thermo Scientific Chemicals

Catalog number: A10269.22
100 g, Each
Thermo Scientific Chemicals

5-Chloro-2-pentanone, 94%, Thermo Scientific Chemicals

Catalog number: A10269.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10269.22
also known as A10269-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5891-21-4
IUPAC Name
5-chloropentan-2-one
Molecular Formula
C5H9ClO
InChI Key
XVRIEWDDMODMGA-UHFFFAOYSA-N
SMILES
CC(=O)CCCCl
Assay (GC)
≥92.0%
Refractive Index
1.4355-1.4425 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to yellow to dark brown to brown
Identification (FTIR)
Conforms

Description

5-Chloro-2-pentanone used in the preparation of destructible surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-pentanone used in the preparation of destructible surfactants.

Solubility
Soluble in chloroform and methanol.

Notes
keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text