Thermo Scientific Chemicals

Trimethyl orthobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A10270.18
50 g, Each
Thermo Scientific Chemicals

Trimethyl orthobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A10270.18
50 g, Each
Quantity
Catalog number: A10270.18
also known as A10270-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
707-07-3
IUPAC Name
(trimethoxymethyl)benzene
Molecular Formula
C10H14O3
InChI Key
IECKAVQTURBPON-UHFFFAOYSA-N
SMILES
COC(OC)(OC)C1=CC=CC=C1
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4875-1.4915 @ 20?C

Description

Trimethyl orthobenzoate was used in the preparation of 5-phenyl-4,6-dipyrrin and 2,3-orthoester derivatives of ethyl-1-thio-α-L-rhamnopyranoside. It reacts with indole to prepare indol-3-yl-phenyl-methanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl orthobenzoate was used in the preparation of 5-phenyl-4,6-dipyrrin and 2,3-orthoester derivatives of ethyl-1-thio-α-L-rhamnopyranoside. It reacts with indole to prepare indol-3-yl-phenyl-methanone.

Solubility
Soluble in organic solvents.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas. Incompatible with oxidizing agents and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text