Thermo Scientific Chemicals

2,5-Difluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A10272.09
10 g, Each
Thermo Scientific Chemicals

2,5-Difluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A10272.09
10 g, Each
Quantity
Catalog number: A10272.09
also known as A10272-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
367-30-6
IUPAC Name
2,5-difluoroaniline
Molecular Formula
C6H5F2N
InChI Key
YNOOQIUSYGWMSS-UHFFFAOYSA-N
SMILES
NC1=CC(F)=CC=C1F
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to yellow to pale brown
Refractive Index
1.5110-1.5150 @ 20?C
Form
Liquid

Description

2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text