Thermo Scientific Chemicals

1-Naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10273.14
25 g, Each
Thermo Scientific Chemicals

1-Naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10273.14
25 g, Each
Quantity
Catalog number: A10273.14
also known as A10273-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-55-5
IUPAC Name
naphthalene-1-carboxylic acid
Molecular Formula
C11H8O2
InChI Key
LNETULKMXZVUST-UHFFFAOYSA-N
SMILES
OC(=O)C1=C2C=CC=CC2=CC=C1
Appearance (Color)
White to cream to brown
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
159.0-165.0°C

Description

1-Naphthoic acid is used as an intermediate for the synthesis of pharmaceuticals, photochemicals, plant growth hormones, dyes and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthoic acid is used as an intermediate for the synthesis of pharmaceuticals, photochemicals, plant growth hormones, dyes and other organic compounds.

Solubility
Freely soluble in hot alcohol and ether. Slightly soluble in hot water.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text