Invitrogen™

Alexa Fluor™ 594 Alkyne (Alexa Fluor™ 594 Carboxamido-(5-(and 6-)Propargyl), Bis(Triethylammonium Salt)), mixed isomers

Catalog number: A10275
Have Questions?
Invitrogen™

Alexa Fluor™ 594 Alkyne (Alexa Fluor™ 594 Carboxamido-(5-(and 6-)Propargyl), Bis(Triethylammonium Salt)), mixed isomers

Catalog number: A10275
Have Questions?
Catalog Number
A10275
Unit Size
0.5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A102750.5 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The red-fluorescent Alexa Fluor® 594 alkyne can be reacted with azides via a copper-catalyzed click reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Solid
Quantity
0.5 mg
Product Type
Alkyne
Molecular Weight (g/mol)
962.63 Da
Solubility
DMSO (Dimethylsulfoxide)
Chemical Reactivity
Azide
Detection Method
Fluorescent
Reactive Group
Alkyne
Shipping Condition
Room Temperature
Reactive Moiety
Alkyne
Label or Dye
Alexa Fluor™ 594
Label Type
Alexa Fluor
Color
Red
Product Line
Alexa Fluor™, Molecular Probes™

Contents & Storage

Store at ≤-20°C, desiccate and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text