Thermo Scientific Chemicals

2-Bromoethanol, 97%, Thermo Scientific Chemicals

Catalog number: A10275.30
250 g, Each
Thermo Scientific Chemicals

2-Bromoethanol, 97%, Thermo Scientific Chemicals

Catalog number: A10275.30
250 g, Each
Quantity
Catalog number: A10275.30
also known as A10275-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
540-51-2
IUPAC Name
2-bromoethan-1-ol
Molecular Formula
C2H5BrO
InChI Key
LDLCZOVUSADOIV-UHFFFAOYSA-N
SMILES
OCCBr
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4900-1.4965 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%

Description

2-Bromoethanol is utilized in selective reduction of nitroarenes. It is used as a solvent and organic synthetic raw material. It finds application in the synthesis of 2-piperidin-1-yl-ethanol by reacting with piperidine. It is used in selective reduction of nitroarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethanol is utilized in selective reduction of nitroarenes. It is used as a solvent and organic synthetic raw material. It finds application in the synthesis of 2-piperidin-1-yl-ethanol by reacting with piperidine. It is used in selective reduction of nitroarenes.

Solubility
Miscible with water, organic solvent and petroleum ether.

Notes
Light sensitive. Hygroscopic. Store in a cool place. Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text