Thermo Scientific Chemicals

Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%, Thermo Scientific Chemicals

Catalog number: A10276.18
50 g, Each
Thermo Scientific Chemicals

Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%, Thermo Scientific Chemicals

Catalog number: A10276.18
50 g, Each
Quantity
Catalog number: A10276.18
also known as A10276-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
334024-15-6
IUPAC Name
iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Formula
C6H5FeO7
InChI Key
NPFOYSMITVOQOS-UHFFFAOYSA-K
SMILES
[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Appearance (Color)
Orange to brown
Form
Powder
Assay from Suppliers CofA
Fe(II) content ≤5.0%
Assay from Suppliers CofA
Fe(III) content ≥16.5% to ≤20.0%
Water Content (Karl Fischer Titration)
≤27.0%

Description

Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

Solubility
Soluble in water.

Notes
Light sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text