3-Chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
3-Chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A10283.06
also known as A10283-06
Price (USD)/ Each
107.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
107.00
Add to cart
3-Chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA10283.06
Price (USD)/ Each
107.00
-
Add to cart
Chemical Identifiers
CAS2888-06-4
IUPAC Name3-chlorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4Cl2O2S
InChI KeyOINWZUJVEXUHCC-UHFFFAOYSA-N
SMILESClC1=CC=CC(=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormFused solid or clear liquid as melt
Appearance (Color)Colorless to pale yellow
Refractive Index1.5665-1.5705 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only