Tri-n-propylamine, 98%, Thermo Scientific Chemicals
Tri-n-propylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-n-propylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number A10285.AK
also known as A10285-AK
Price (USD)/ Each
28.65
Online exclusive
31.50 
Save 2.85 (9%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
28.65
Online exclusive
31.50 
Save 2.85 (9%)
Add to cart
Tri-n-propylamine, 98%, Thermo Scientific Chemicals
Catalog numberA10285.AK
Price (USD)/ Each
28.65
Online exclusive
31.50 
Save 2.85 (9%)
-
Add to cart
Chemical Identifiers
CAS102-69-2
IUPAC Nametripropylamine
Molecular FormulaC9H21N
InChI KeyYFTHZRPMJXBUME-UHFFFAOYSA-N
SMILESCCCN(CCC)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤1%
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
View more

Tri-n-propylamine is used as an chemical intermediate, organic solvent. It is used in the preparation of chemical compounds such as dyes, drugs, catalyst & other uses. It is used together with ruthenium in the electrochemiluminescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-propylamine is used as an chemical intermediate, organic solvent. It is used in the preparation of chemical compounds such as dyes, drugs, catalyst & other uses. It is used together with ruthenium in the electrochemiluminescence.

Solubility
Soluble in water (2.6 g/L) at 20°C, ether and ethanol.

Notes
Hygroscopic. Incompatible with oxidizing agents, acids.
RUO – Research Use Only
Rebecca Y. Lai and Allen J. Bard. Electrogenerated Chemiluminescence. 70. The Application of ECL to Determine Electrode Potentials of Tri-n-propylamine, Its Radical Cation, and Intermediate Free Radical in MeCN/Benzene Solutions. J. Phys. Chem. A.2003, 107 (18), 3335-3340.