4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.
Solubility
Soluble in water.
Notes
Incompatible with strong acids, strong bases and strong oxidizing agents.
RUO – Research Use Only
Naika, R. M.; Kumar, B.; Prasad, S.; Chetty, A. A.; Asthana, A. Catalytic ligand exchange reaction between hexacyanoferrate(II) and 4-cyanopyridine applied to trace kinetic analysis of palladium(II). Microchem. J. 2015, 122, 82-88.
Naik, R. M.; Asthana, A.; Yadav, S. B. S. Comparative study of the catalytic effect of Ag(I) and Hg(II) on the exchange of cyanide by 4-cyanopyridine in hexacyanoferrate(II): a kinetic approach. J. Iran. Chem. Soc. 2015, 12 (3), 519-528.