o-Anisidine, 99%, Thermo Scientific Chemicals
o-Anisidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Anisidine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10288.30
also known as A10288-30
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.60 
Save 5.95 (14%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.60 
Save 5.95 (14%)
Add to cart
o-Anisidine, 99%, Thermo Scientific Chemicals
Catalog numberA10288.30
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.60 
Save 5.95 (14%)
-
Add to cart
Chemical Identifiers
CAS90-04-0
IUPAC Name2-methoxyaniline
Molecular FormulaC7H9NO
InChI KeyVMPITZXILSNTON-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to red or brown
Assay (GC)≥98.5%
Refractive Index1.5735-1.5765 @ 20?C
FormLiquid
Identification (FTIR)Conforms
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Andrzej Sapota; Bogdan Czerski; Marian Jedrzejczak. Tissue distribution, excretion and metabolism of o-anisidine in rats. International Journal of Occupational Medicine and Environmental Health. 2003, 16 (4), 351-357.