Thermo Scientific Chemicals

3-(4-Methylbenzoyl)propionic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10290.14
25 g, Each
Thermo Scientific Chemicals

3-(4-Methylbenzoyl)propionic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10290.14
25 g, Each
Quantity
Catalog number: A10290.14
also known as A10290-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4619-20-9
IUPAC Name
4-(4-methylphenyl)-4-oxobutanoic acid
Molecular Formula
C11H12O3
InChI Key
OEEUWZITKKSXAZ-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C(=O)CCC(O)=O
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
123.0-130.0?C
Form
Crystals or powder or crystalline powder

Description

3-(4-Methylbenzoyl)propionic acid can be used as an intermediate in agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Methylbenzoyl)propionic acid can be used as an intermediate in agrochemical, pharmaceutical and dyestuff field

Solubility
Insoluble in water.

Notes
Incompatible with bases and oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text